missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane 97.0+%, TCI America™

Catalog No. p-7136061
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T006325G 25 g
T0063500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T006325G Supplier TCI America Supplier No. T006325G
Only null left

Chemical Identifiers

CAS 79-34-5
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.838
MDL Number MFCD00000848
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
Synonym s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
PubChem CID 6591
ChEBI CHEBI:36026
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Melting Point -44°C
Color Yellow
Boiling Point 146°C
Quantity 25 g
UN Number 1702
Formula Weight 167.84
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrachloroethane
EINECSNumber (2)-0056
RTECSNumber KI8575000
TSCA Yes