missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane, (TCE), Reagents

Catalog No. 01336935
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336935 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336935 Supplier Labchem Inc Supplier No. C23374001Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1,1,2,2-Tetrachloroethane, (TCE), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 79-34-5
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.84
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
Synonym TCE, s-Tetrachloroethane, Acetylene tetrachloride
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES ClC(Cl)C(Cl)Cl

Specifications

Boiling Range 146.5°C
Melting Point -44°C
Color Colorless to Slight Yellow
Boiling Point 146.5°C
Melting Range -44°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.595
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrachloroethane, (TCE)
Show More Show Less