missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane, 98+%

Catalog No. p-7044090
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0448914 25 g
AAL0448922 100 g
AAL0448936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL0448914 Supplier Thermo Scientific Chemicals Supplier No. L0448914
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 79-34-5 | C2H2Cl4 | 167.838 g/mol

1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.

Solubility
Miscible with water, methanol, ethanol, chloroform and acetone.

Notes
Incompatible with strong oxidizing agents, sodium, sodium oxides, strong bases, potassium, nitrates, 2,4-dinitrophenyl disulfide.

Chemical Identifiers

CAS 79-34-5
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.838
MDL Number MFCD00000848
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
Synonym s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
PubChem CID 6591
ChEBI CHEBI:36026
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Melting Point -43°C
Density 1.596
Boiling Point 142°C to 146°C
Refractive Index 1.4935
Linear Formula Cl2CHCHCl2
Quantity 25 g
UN Number UN1702
Beilstein 969206
Merck Index 14,9189
Solubility Information Miscible with water,methanol,ethanol,chloroform and acetone.
Formula Weight 167.85
Percent Purity ≥98%
Chemical Name or Material 1,1,2,2-Tetrachloroethane
Show More Show Less
Hazard Category H310+H330-H351
Hazard Statement GHS H Statement
H310-H330-H341-H351
Fatal in contact with skin.
Fatal if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P262-P264b-P270-P271-P281-P284-P302+P350-P304+P340-P308+P313-P310-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,1,2,2-TETRACHLOROETHANE
EINECSNumber 201-197-8
RTECSNumber KI8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only