missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane, 98%, Spectrum™ Chemical

Catalog No. 18600019
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600019 250 mL Amber Glass Bottle
18600021 1 L Amber Glass Bottle
18600022 4 L Amber Glass Bottle
18600020 20 L Steel Can
18600023 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600019 Supplier Spectrum Chemical Mfg Cor Supplier No. T1018250ML
Get Up to $500 in Chemicals with Purchase
View all promotions

T1018, 79-34-5, C2H2Cl4

Spectrum™ Chemical 1,1,2,2-Tetrachloroethane , also known as Acetylene tetrachloride, is a chlorinated derivative of ethane with a very high solvent power. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 79-34-5
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.84
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES ClC(Cl)C(Cl)Cl

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 250 mL
UN Number UN1702
Percent Purity 98%
Grade Reagent