missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2-Tetrachloro-1,2-dimethyldisilane 98.0+%, TCI America™
$382.13 - $1288.06
Chemical Identifiers
| CAS | 4518-98-3 |
|---|---|
| Molecular Formula | C2H6Cl4Si2 |
| Molecular Weight (g/mol) | 228.04 |
| MDL Number | MFCD00192469 |
| InChI Key | JTBAMRDUGCDKMS-UHFFFAOYSA-N |
| PubChem CID | 78276 |
| IUPAC Name | 1,1,2,2-tetrachloro-1,2-dimethyldisilane |
| SMILES | C[Si](Cl)(Cl)[Si](C)(Cl)Cl |
Chemical Identifiers
| 4518-98-3 | |
| 228.04 | |
| JTBAMRDUGCDKMS-UHFFFAOYSA-N | |
| 1,1,2,2-tetrachloro-1,2-dimethyldisilane |
| C2H6Cl4Si2 | |
| MFCD00192469 | |
| 78276 | |
| C[Si](Cl)(Cl)[Si](C)(Cl)Cl |
Specifications
| 4518-98-3 | |
| C2H6Cl4Si2 | |
| 5 g | |
| JTBAMRDUGCDKMS-UHFFFAOYSA-N | |
| 1,1,2,2-tetrachloro-1,2-dimethyldisilane | |
| 78276 | |
| ≥98.0% (T) | |
| 1,1,2,2-Tetrachloro-1,2-dimethyldisilane |
| Colorless | |
| MFCD00192469 | |
| 2988 | |
| C[Si](Cl)(Cl)[Si](C)(Cl)Cl | |
| 228.04 | |
| 228.04 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2040
TSCA : Yes
Recommended Storage : Refrigerator