missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane 97.0+%, TCI America™

Catalog No. p-7131130
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16105G 5 g
H161025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16105G Supplier TCI America Supplier No. H16105G
Only null left

Chemical Identifiers

CAS 40573-09-9
Molecular Formula C8F16O5
Molecular Weight (g/mol) 480.06
MDL Number MFCD00506884
InChI Key HCGYVDVMTQESCQ-UHFFFAOYSA-N
PubChem CID 57349774
IUPAC Name hexadecafluoro-2,4,6,8,11-pentaoxatridec-12-ene
SMILES FC(F)=C(F)OC(F)(F)C(F)(F)OC(F)(F)OC(F)(F)OC(F)(F)OC(F)(F)F

Specifications

Color Colorless
Boiling Point 64°C
Quantity 5 g
Formula Weight 332.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane
TSCA No
Recommended Storage Refrigerator