missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2,3,3,4-Heptafluorocyclopentane 98.0+%, TCI America™

Catalog No. H1013500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1013500G 500 g
H101325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1013500G Supplier TCI America Supplier No. H1013500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15290-77-4
Molecular Formula C5H3F7
Molecular Weight (g/mol) 196.068
MDL Number MFCD03844796
InChI Key IDBYQQQHBYGLEQ-UHFFFAOYSA-N
PubChem CID 3853245
IUPAC Name 1,1,2,2,3,3,4-heptafluorocyclopentane
SMILES C1C(C(C(C1(F)F)(F)F)(F)F)F

Specifications

Color White
Boiling Point 83°C
Quantity 500 g
Formula Weight 196.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,1,2,2,3,3,4-Heptafluorocyclopentane
EINECSNumber (3)-4446
TSCA Yes