missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoroacetone cyanohydrin, 95%

Catalog No. AAL1131814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1131814 25 g
AAL1131806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1131814 Supplier Thermo Scientific Chemicals Supplier No. L1131814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 335-08-0 | C4H4F3NO | 139.077 g/mol

A chlorinated acetone derivative for organic synthesis and proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chlorinated acetone derivative for organic synthesis and proteomics research

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.

Chemical Identifiers

CAS 335-08-0
Molecular Formula C4H4F3NO
Molecular Weight (g/mol) 139.077
MDL Number MFCD00040885
InChI Key XDCMNDCKYSQKAX-UHFFFAOYSA-N
Synonym 1,1,1-trifluoroacetone cyanohydrin, 2-cyano-1,1,1-trifluoro-2-propanol, 2-hydroxy-2-trifluoromethyl propionitrile, propanenitrile, 3,3,3-trifluoro-2-hydroxy-2-methyl, 2-trifluoromethyl lactonitrile, trifluoroacetone cyanohydrin, lactonitrile, 2-trifluoromethyl, 3-trifluoro-a-hydroxyisobutyronitrile, lactonitrile,3,3-trifluoro-2-methyl, 2-cyano-1,1,1-trifluoropropan-2-ol
PubChem CID 274400
IUPAC Name 3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile
SMILES CC(C#N)(C(F)(F)F)O

Specifications

Density 1.276
Boiling Point 68°C to 69°C (40 mmHg)
Flash Point 60°C (140°F)
Refractive Index 1.338
Quantity 25 g
UN Number UN2929
Beilstein 1758723
Sensitivity Moisture sensitive
Solubility Information Fully miscible in water.
Formula Weight 139.08
Percent Purity 95%
Chemical Name or Material 1,1,1-Trifluoroacetone cyanohydrin
Show More Show Less
Hazard Category H226-H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only