missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoroacetone, 95%

CAS: 421-50-1 | C3H3F3O | 112.05 g/mol

$232.71 - $232.71

Chemical Identifiers

CAS 421-50-1
Molecular Formula C3H3F3O
Molecular Weight (g/mol) 112.05
InChI Key FHUDAMLDXFJHJE-UHFFFAOYSA-N
Synonym 1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone
PubChem CID 9871
IUPAC Name 1,1,1-trifluoropropan-2-one
SMILES CC(=O)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC139740250
SDS View Documents
Thermo Scientific Chemicals
139740250
25 g Glass bottle
Each for $232.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

421-50-1
112.05
1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone
1,1,1-trifluoropropan-2-one
C3H3F3O
FHUDAMLDXFJHJE-UHFFFAOYSA-N
9871
CC(=O)C(F)(F)F

Specifications

421-50-1
Colorless
22.0°C
Authentic
Glass bottle
1.3000 (20°C, 589nm) max.
25 g
13,323; 15,212
1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone
FHUDAMLDXFJHJE-UHFFFAOYSA-N
1,1,1-trifluoropropan-2-one
9871
112.05
Liquid
-78.0°C
1.2500g/mL
−30°C
98% min. (GC)
C3H3F3O
CF3COCH3
01,II,717
1.25
Solubility in water: miscible
CC(=O)C(F)(F)F
112.05
3.28 mPa.s (15°C)
98+%
1, 1, 1-Trifluoroacetone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 207-005-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only