missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-3-phenylacetone, 97%

Catalog No. AAL1685103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1685103 1 g
AAL1685106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1685103 Supplier Thermo Scientific Chemicals Supplier No. L1685103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 350-92-5 | C9H7F3O | 188.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 350-92-5
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00068171
InChI Key IAJKTOIWQHTZOS-UHFFFAOYSA-N
Synonym 3-phenyl-1,1,1-trifluoropropan-2-one, 1,1,1-trifluoro-3-phenylacetone, 1,1,1-trifluoro-3-phenyl-2-propanone, 1,1,1-trifluoro-3-phenyl-propan-2-one, 2-propanone, 1,1,1-trifluoro-3-phenyl, acmc-20amz1, trifluoromethylbenzyl ketone, benzyl trifluoromethyl ketone, trifluoromethyl benzyl ketone, ksc227m3d
PubChem CID 222958
IUPAC Name 1,1,1-trifluoro-3-phenylpropan-2-one
SMILES C1=CC=C(C=C1)CC(=O)C(F)(F)F

Specifications

Density 1.22
Boiling Point 51°C to 52°C (2 mmHg)
Refractive Index 1.447
Quantity 1 g
Beilstein 2614467
Formula Weight 188.15
Percent Purity 97%
Chemical Name or Material 1,1,1-Trifluoro-3-phenylacetone
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only