missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate 98.0+%, TCI America™

Catalog No. p-7136251
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
T038625G 25 g
T0386500G 500 g
2 options

Catalog No. T038625G

Supplier: TCI America T038625G

Only null left

Chemical Identifiers

CAS 6001-64-5
Molecular Formula C4H11Cl3O2
Molecular Weight (g/mol) 197.48
MDL Number MFCD00004461
InChI Key HBARVHNVVKZUGS-UHFFFAOYSA-N
Synonym c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate
PubChem CID 102594540
IUPAC Name molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate
SMILES [HH].CC(C)(C(Cl)(Cl)Cl)O.O

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 175°C
Quantity 25 g
Formula Weight 177.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate
EINECSNumber (2)-2002
TSCA Yes