missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%

CAS: 6001-64-5 | 0·5 H2O | 186.47 g/mol

$281.94 - $281.94

Chemical Identifiers

CAS 6001-64-5
Molecular Formula 0·5 H2O
Molecular Weight (g/mol) 186.47
MDL Number MFCD02179352
InChI Key HBARVHNVVKZUGS-UHFFFAOYSA-N
Synonym c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate
PubChem CID 102594540
IUPAC Name molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate
SMILES [HH].CC(C)(C(Cl)(Cl)Cl)O.O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC150305000
SDS View Documents
Thermo Scientific Chemicals
150305000
500 g Plastic bottle
Each for $281.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6001-64-5
186.47
HBARVHNVVKZUGS-UHFFFAOYSA-N
102594540
[HH].CC(C)(C(Cl)(Cl)Cl)O.O
0·5 H2O
MFCD02179352
c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate
molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate

Specifications

6001-64-5
100.0
White
100°C
97.5 to 102.5% (ex Cl) (Argentometry)
0·5 H2O
MFCD02179352
c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate
HBARVHNVVKZUGS-UHFFFAOYSA-N
molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate
102594540
98%
1, 1, 1-Trichloro-2-methyl-2-propanol hemihydrate, 98%
97.5
75°C to 79°C
173°C to 175°C
Authentic
Plastic bottle
0·5H2O
500 g
Solubility in water: 7.7g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,glycerol
[HH].CC(C)(C(Cl)(Cl)Cl)O.O
186.47
186.47
Crystalline Powder, Crystals or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only