missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane 95.0+%, TCI America™
Supplier: TCI America O039725G
Specifications
| 1,1,1,3,5,7,7,7-Octamethyltetrasiloxane | |
| Colorless | |
| C8H24O3Si4 | |
| 25 g | |
| OHSYWAVRSCQMHG-UHFFFAOYSA-N | |
| methyl-[methyl(trimethylsilyloxy)-$l^{3}-silanyl]oxy-trimethylsilyloxysilicon | |
| 6327318 | |
| ≥95.0% (GC) |
| 16066-09-4 | |
| 177°C | |
| MFCD00053664 | |
| 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, 3h,5h-octamethyltetrasiloxane, 1,3-bis trimethylsiloxy-1,3-dimethyldisiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl, bis trimethylsiloxy dimethyldisiloxane, methyl-methyl trimethylsilyloxy, 1,1,1,3,5,7,7,7-octamethyl-tetrasiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl-,, 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, technical gc | |
| C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C | |
| 280.617 | |
| 282.63 | |
| Liquid |
Chemical Identifiers
| 16066-09-4 | |
| 280.617 | |
| OHSYWAVRSCQMHG-UHFFFAOYSA-N | |
| 6327318 | |
| C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C |
| C8H24O3Si4 | |
| MFCD00053664 | |
| 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, 3h,5h-octamethyltetrasiloxane, 1,3-bis trimethylsiloxy-1,3-dimethyldisiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl, bis trimethylsiloxy dimethyldisiloxane, methyl-methyl trimethylsilyloxy, 1,1,1,3,5,7,7,7-octamethyl-tetrasiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl-,, 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, technical gc | |
| methyl-[methyl(trimethylsilyloxy)-$l^{3}-silanyl]oxy-trimethylsilyloxysilicon |
Safety and Handling
EINECSNumber : (7)-0477
TSCA : Yes