missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,5,7,7,7-Octamethyltetrasiloxane 95.0+%, TCI America™

Catalog No. O03975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03975G 5 g
O039725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03975G Supplier TCI America Supplier No. O03975G

Chemical Identifiers

CAS 16066-09-4
Molecular Formula C8H24O3Si4
Molecular Weight (g/mol) 280.617
MDL Number MFCD00053664
InChI Key OHSYWAVRSCQMHG-UHFFFAOYSA-N
Synonym 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, 3h,5h-octamethyltetrasiloxane, 1,3-bis trimethylsiloxy-1,3-dimethyldisiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl, bis trimethylsiloxy dimethyldisiloxane, methyl-methyl trimethylsilyloxy, 1,1,1,3,5,7,7,7-octamethyl-tetrasiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl-,, 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, technical gc
PubChem CID 6327318
IUPAC Name methyl-[methyl(trimethylsilyloxy)-$l^{3}-silanyl]oxy-trimethylsilyloxysilicon
SMILES C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 177°C
Quantity 5 g
Formula Weight 282.63
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,5,7,7,7-Octamethyltetrasiloxane
EINECSNumber (7)-0477
TSCA Yes