missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,5,5,5-Heptamethyltrisiloxane 98.0+%, TCI America™

Catalog No. h126725ml
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H126725ML 25 mL
H1267250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H126725ML Supplier TCI America Supplier No. H126725ML
Only null left

Chemical Identifiers

CAS 1873-88-7
Molecular Formula C7H21O2Si3
Molecular Weight (g/mol) 221.498
MDL Number MFCD00048000
InChI Key SWGZAKPJNWCPRY-UHFFFAOYSA-N
Synonym 1,1,1,3,5,5,5-heptamethyltrisiloxane, bis trimethylsiloxy methylsilane, unii-rvy9659r8c, trisiloxane, 1,1,1,3,5,5,5-heptamethyl, methyl-bis trimethylsilyloxy silicon, dsstox_cid_18795, methicone 20 cst, unii-6777u11mkt, methylpolysilicone, xiameter mhx-1107 fluid 20 cst
PubChem CID 6327366
IUPAC Name methyl-bis(trimethylsilyloxy)silicon
SMILES C[Si](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 142°C
Quantity 25 mL
UN Number 1993
Formula Weight 222.51
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,5,5,5-Heptamethyltrisiloxane