missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexamethyldisilazane 96.0+%, TCI America™

Catalog No. H008925ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H008925ML 25 mL
H0089100ML 100 mL
H0089500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H008925ML Supplier TCI America Supplier No. H008925ML

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
Molecular Weight (g/mol) 161.395
MDL Number MFCD00008259
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 mL
UN Number 3286
Formula Weight 161.40
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexamethyldisilazane
EINECSNumber (2)-2044&(2)-2955&(9)-1324
RTECSNumber JM9230000
TSCA Yes