missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,3,3-Hexamethyldisilazane, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -78.0°C |
| Color | Colorless |
| Density | 0.7600g/mL |
| Boiling Point | 125.0°C |
| Flash Point | 20°C |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4060 to 1.4090 |
| Linear Formula | (CH3)3SiNHSi(CH3)3 |
| Quantity | 10 mL |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. Toxic in contact with skin. Harmful if inhaled. Harmful to aquatic life with long lasting effects. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with |
| EINECSNumber | 213-668-5 |
RUO – Research Use Only