missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexamethyldisilazane, 98%, AcroSeal™

CAS: 999-97-3 | C6H19NSi2 | 161.4 g/mol

Supplier:  Thermo Scientific Chemicals 430850010

 View more versions of this product

Catalog No. AC430850010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
Specifications

Specifications

1,1,1,3,3,3-Hexamethyldisilazane
97.5
-78.0°C
125.0°C
Authentic
AcroSeal™ Glass Bottle
1.4060 to 1.4090
MFCD00008259
3286
15,4725
Solubility in water: reacts. Other solubilities: reacts with methanol, soluble in acetone, benzene, ethyl ether, pentane, and perchloroethylene
C[Si](C)(C)N[Si](C)(C)C
161.4
0.9 mm2/s (25°C)
161.4
Liquid
999-97-3
100.0
0.7600g/mL
20°C
97.5% min. (GC)
C6H19NSi2
(CH3)3SiNHSi(CH3)3
1 L
0.76
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
FFUAGWLWBBFQJT-UHFFFAOYSA-N
[dimethyl-(trimethylsilylamino)silyl]methane
13838
CHEBI:85068
98%

Chemical Identifiers

999-97-3
161.4
FFUAGWLWBBFQJT-UHFFFAOYSA-N
13838
[dimethyl-(trimethylsilylamino)silyl]methane
C6H19NSi2
MFCD00008259
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
CHEBI:85068
C[Si](C)(C)N[Si](C)(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

EINECSNumber : 213-668-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only