missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl Trifluoromethanesulfonate 97.0+%, TCI America™

Catalog No. H12765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12765G Supplier TCI America Supplier No. H12765G
Only null left

Chemical Identifiers

CAS 156241-41-7
Molecular Formula C4HF9O3S
Molecular Weight (g/mol) 300.095
MDL Number MFCD02093340
InChI Key NRHQWNHARTXNOE-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 1,1,1,3,3,3-Hexafluoroisopropyl Ester, 1,1,1,3,3,3-Hexafluoroisopropyl Triflate
PubChem CID 2775011
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl trifluoromethanesulfonate
SMILES C(C(F)(F)F)(C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
UN Number 1760
Formula Weight 300.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl Trifluoromethanesulfonate