missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. M098825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M098825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M098825G Supplier TCI America Supplier No. M098825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 3063-94-3
Color Colorless
Molecular Formula C7H6F6O2
MDL Number MFCD00040105
Quantity 25 g
UN Number 3272
Synonym hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma
InChI Key FMQPBWHSNCRVQJ-UHFFFAOYSA-N
SMILES CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
Molecular Weight (g/mol) 236.113
PubChem CID 76469
Formula Weight 236.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl Methacrylate (stabilized with MEHQ)
Show More Show Less
EINECSNumber (2)-1031
TSCA Yes