Learn More
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, 99%, stab.
CAS: 3063-94-3 | C7H6F6O2 | 236.113 g/mol
$90.19 - $328.34
Chemical Identifiers
| CAS | 3063-94-3 |
|---|---|
| Molecular Formula | C7H6F6O2 |
| Molecular Weight (g/mol) | 236.113 |
| MDL Number | MFCD00040105 |
| InChI Key | FMQPBWHSNCRVQJ-UHFFFAOYSA-N |
| Synonym | hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma |
| PubChem CID | 76469 |
| IUPAC Name | 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2144706
|
Thermo Scientific Chemicals
B2144706 |
5 g |
Each for $90.19
|
|
|||||
|
AAB2144714
|
Thermo Scientific Chemicals
B2144714 |
25 g |
Each for $328.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3063-94-3 | |
| 236.113 | |
| FMQPBWHSNCRVQJ-UHFFFAOYSA-N | |
| 76469 | |
| CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F |
| C7H6F6O2 | |
| MFCD00040105 | |
| hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma | |
| 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate |
Specifications
| 3063-94-3 | |
| 99°C | |
| C7H6F6O2 | |
| MFCD00040105 | |
| UN1993 | |
| FMQPBWHSNCRVQJ-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate | |
| 76469 | |
| 99% |
| 1.302 | |
| 14°C (57°F) | |
| 1.331 | |
| 5 g | |
| hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma | |
| CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F | |
| 236.113 | |
| 236.12 | |
| 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, stabilized |
Safety and Handling
GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H224-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 221-309-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only