Learn More
1,1,1,3,3,3-Hexafluoroisopropyl acrylate, 98+%, stab. with 50ppm 4-methoxyphenol
Used in agrochemical, pharmaceutical and dyestuff field. | CAS: 2160-89-6 | C6H4F6O2 | 222.09 g/mol
$474.86 - $474.86
Chemical Identifiers
| CAS | 2160-89-6 |
|---|---|
| Molecular Formula | C6H4F6O2 |
| Molecular Weight (g/mol) | 222.09 |
| MDL Number | MFCD00040104 |
| InChI Key | MNSWITGNWZSAMC-UHFFFAOYSA-N |
| Synonym | 1,1,1,3,3,3-hexafluoroisopropyl acrylate, hexafluoroisopropyl acrylate, 1,1,1,3,3,3-hexafluoropropan-2-yl acrylate, 2-propenoic acid, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, hexafluoro-2-propyl acrylate, mnswitgnwzsamc-uhfffaoysa, 1h-1-trifluoromethyl trifluoroethyl acrylate |
| PubChem CID | 75096 |
| IUPAC Name | 1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate |
| SMILES | FC(F)(F)C(OC(=O)C=C)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1212814
|
Thermo Scientific Chemicals
L1212814 |
25 g |
Each for $474.86
|
|
|||||
Description
1,1,1,3,3,3-Hexafluoroisopropyl acrylate is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2160-89-6 | |
| 222.09 | |
| MNSWITGNWZSAMC-UHFFFAOYSA-N | |
| 75096 | |
| FC(F)(F)C(OC(=O)C=C)C(F)(F)F |
| C6H4F6O2 | |
| MFCD00040104 | |
| 1,1,1,3,3,3-hexafluoroisopropyl acrylate, hexafluoroisopropyl acrylate, 1,1,1,3,3,3-hexafluoropropan-2-yl acrylate, 2-propenoic acid, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, hexafluoro-2-propyl acrylate, mnswitgnwzsamc-uhfffaoysa, 1h-1-trifluoromethyl trifluoroethyl acrylate | |
| 1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate |
Specifications
| 2160-89-6 | |
| 1.37 | |
| 17°C (62°F) | |
| C6H4F6O2 | |
| H2C=CHCO2CH(CF3)2 | |
| 25 g | |
| 2094742 | |
| 1,1,1,3,3,3-hexafluoroisopropyl acrylate, hexafluoroisopropyl acrylate, 1,1,1,3,3,3-hexafluoropropan-2-yl acrylate, 2-propenoic acid, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, hexafluoro-2-propyl acrylate, mnswitgnwzsamc-uhfffaoysa, 1h-1-trifluoromethyl trifluoroethyl acrylate | |
| MNSWITGNWZSAMC-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate | |
| 75096 | |
| ≥98% | |
| 1,1,1,3,3,3-Hexafluoroisopropyl acrylate |
| Colorless | |
| 81°C to 82°C | |
| Glass bottle | |
| 1.319 | |
| MFCD00040104 | |
| UN3272 | |
| 1.37 | |
| Not miscible in water. | |
| FC(F)(F)C(OC(=O)C=C)C(F)(F)F | |
| 222.09 | |
| 222.09 | |
| Liquid |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 218-479-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only