missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,3,3-Hexafluoro-2-propanol 99.0+%, TCI America™
Supplier: TCI America H0424100G
Specifications
| 1,1,1,3,3,3-Hexafluoro-2-propanol | |
| -4°C | |
| 58°C | |
| MFCD00011651 | |
| 1760 | |
| BYEAHWXPCBROCE-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoropropan-2-ol | |
| 13529 | |
| 168.04 | |
| Liquid |
| 920-66-1 | |
| Colorless | |
| C3H2F6O | |
| 100 g | |
| 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
| C(C(F)(F)F)(C(F)(F)F)O | |
| 168.038 | |
| CHEBI:63104 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 920-66-1 | |
| 168.038 | |
| BYEAHWXPCBROCE-UHFFFAOYSA-N | |
| 13529 | |
| 1,1,1,3,3,3-hexafluoropropan-2-ol |
| C3H2F6O | |
| MFCD00011651 | |
| 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
| CHEBI:63104 | |
| C(C(F)(F)F)(C(F)(F)F)O |
Safety and Handling
EINECSNumber : (2)-0291
RTECSNumber : UB6450000
TSCA : Yes