missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, Reagent, 99%, Spectrum™ Chemical

Catalog No. p-7157531
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600014 5 g Amber Glass Bottle
18600015 25 g Amber Glass Bottle
18600016 100 g Amber Glass Bottle
18600017 1 kg Amber Glass Bottle
18600018 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600014 Supplier Spectrum Chemical Mfg Cor Supplier No. H10085GM
Only null left

H1008, 920-66-1, C3H2F6O

Spectrum™ Chemical 1,1,1,3,3,3-Hexafluoro-2-propanol, Reagent , is the organic compound that is commonly abbreviated as HFIP and is a fluorinated alcohol that is used as a solvent and synthetic intermediate. It′s a colorless, volatile liquid that has a characteristic strong, pungent odor. It is mainly used for solubilizing a rangeof polymers as well as being the precursor and chief metabolite of the inhalation anesthetic sevoflurane. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.04
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES OC(C(F)(F)F)C(F)(F)F

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
UN Number UN1760
Percent Purity 99%
Grade Reagent