missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, LC-MS Grade, Honeywell Fluka™

Catalog No. p-7120006
Change view
Click to view available options
Quantity:
50 mL
10 x 1 mL
Packaging:
Ampoule
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046851 10 x 1 mL Ampoule
60046852 50 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046851 Supplier Honeywell Chemicals Supplier No. 4206010X1ML
Only null left

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.038
MDL Number MFCD00011651
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifications

Melting Point -4°C
Color Colorless
Density 1.596g/mL at 25°C
Boiling Point 59°C
Packaging Ampoule
Refractive Index n20/D 1.275
Linear Formula (CF3)2CHOH
Quantity 10 x 1 mL
UN Number UN3265
Beilstein 1841007
Residue after Evaporation 0 to 0.0005%
Formula Weight 168.04g/mol
Percent Purity ≥99.8% (GC)
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 8/PGII
RTECSNumber 213-059-4