missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, MilliporeSigma™

For synthesis

Supplier:  Technidata 8045150100

Catalog No. M8045150100

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,1,1,3,3,3-hexafluoro-2-propanol
3°C
1.62g/cm3 (20°C)
58°C
Characteristic
C3H2F6O
100 mL
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoropropan-2-ol
13529
168.03
Liquid
920-66-1
Colorless
3 to 4 (aq. soln.)
>100°C (>212°F)
Glass Bottle
1.2752 (25°C, 589nm)
UN 3265
Water: soluble at 20°C (68°F)
C(C(F)(F)F)(C(F)(F)F)O
168.038
CHEBI:63104
269 hPa (30°C)

Chemical Identifiers

920-66-1
168.038
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
CHEBI:63104
C(C(F)(F)F)(C(F)(F)F)O
C3H2F6O
BYEAHWXPCBROCE-UHFFFAOYSA-N
13529
1,1,1,3,3,3-hexafluoropropan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H302 + H332: Harmful if swallowed or if inhaled

H314: Causes severe skin burns and eye damage.

H290: May be corrosive to metals.

EINECSNumber : 213-059-4

RTECSNumber : UB6450000

Recommended Storage : 15° to 25°C (59° to 77°F); Tightly closed

SDS
Documents

Documents

Promotions

Promotions