missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy

Catalog No. AC293410100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC293410100 10 g Glass bottle
AC293410500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC293410100 Supplier Thermo Scientific Chemicals Supplier No. 293410100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 920-66-1 | C3H2F6O | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

Chemical Identifiers

CAS 920-66-1
MDL Number MFCD00011651
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifications

Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.009 max. at 240nm, 0.046 max. at 200nm, 0.097 max. at 195nm, 0.301 max. at 190nm
CAS Min % 99.85
CAS Max % 100.0
Melting Point -4.0°C
Density 1.5960g/mL
Boiling Point 59.0°C
Flash Point >100°C
Assay Percent Range 99.85% min. (GC)
Packaging Glass bottle
Molecular Formula C3H2F6O
Refractive Index 1.2740 to 1.2770
Linear Formula (CF3)2CHOH
Quantity 10 g
Residue after Evaporation 0.001% max.
Specific Gravity 1.596
Solubility Information Solubility in water: 1000g/L (25°C)
Molecular Weight (g/mol) 168.04
Viscosity 1.65 mPa.s (20°C)
Formula Weight 168.04
Percent Purity 99.9%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 213-059-4

RUO – Research Use Only