missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1,1,1,3,3,3-Hexafluoro-2-Propanol

Catalog No. 1052285G
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1052285G 5 g
105228100G 100 g
1052282KG 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1052285G Supplier Sigma Aldrich Supplier No. 1052285G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,1,1,3,3,3-Hexafluoro-2-Propanol

Specifications

CAS 920-66-1
Melting Point -4°C
Density 1.596 g/mL at 25°C
Boiling Point 59°C
Molecular Formula C3H2F6O
Linear Formula (CF3)2CHOH
Refractive Index n20/D 1.275
Quantity 5 g
MDL Number MFCD00011651
Synonym HFP; Hexafluoroisopropanol
Molecular Weight (g/mol) 168.04 g/mol
Show More Show Less