missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone 98.0+%, TCI America™

Catalog No. N103825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N103825G 25 g
N1038100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N103825G Supplier TCI America Supplier No. N103825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 756-13-8
Molecular Formula C6F12O
Molecular Weight (g/mol) 316.05
MDL Number MFCD04038343
InChI Key RMLFHPWPTXWZNJ-UHFFFAOYSA-N
Synonym Perfluoroethyl Perfluoroisopropyl Ketone
PubChem CID 2782408
IUPAC Name 1,1,1,2,2,4,5,5,5-nonafluoro-4-(trifluoromethyl)pentan-3-one
SMILES FC(F)(F)C(F)(F)C(=O)C(F)(C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Quantity 25 g
Formula Weight 316.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone
EINECSNumber (2)-4024
TSCA Yes