missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,2,2,3,3-Heptachloropropane 98.0+%, TCI America™

Catalog No. H001525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H001525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H001525G Supplier TCI America Supplier No. H001525G
Only null left

Chemical Identifiers

CAS 594-89-8
Molecular Formula C3HCl7
Molecular Weight (g/mol) 285.191
MDL Number MFCD00000847
InChI Key YFIIENAGGCUHIQ-UHFFFAOYSA-N
PubChem CID 11677
IUPAC Name 1,1,1,2,2,3,3-heptachloropropane
SMILES C(C(C(Cl)(Cl)Cl)(Cl)Cl)(Cl)Cl

Specifications

Color White-Yellow
Boiling Point 247°C
Quantity 25 g
Formula Weight 285.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,1,1,2,2,3,3-Heptachloropropane