Learn More
Charles River Laboratories Inc 1-.01 EU/ML CARTRIDGES 10/PK
Catalog No. NC9744492
Encompass
10 single packs of cartridges All of our rapid testing instruments utilize our FDA-licensed cartridges for batch release testing, as well as a variety of non-regulated cartridges for simple, 15-minute testing. The cartridges are pre-loaded with all of the reagents required to perform an LAL test, eliminating preparation of multiple reagents and reducing the opportunity for technician error. Since there is no need to create a standard curve, create a positive product control and reconstitute LAL, there is no need for a negative control by the user. However, it is recommended that when performing sample dilutions, the diluent be tested for endotoxin prior to use.
Catalog No. NC9744492 Supplier Charles River Laboratories Inc Supplier No. PTS2001F
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10 single packs of cartridges All of our rapid testing instruments utilize our FDA-licensed cartridges for batch release testing, as well as a variety of non-regulated cartridges for simple, 15-minute testing. The cartridges are pre-loaded with all of the reagents required to perform an LAL test, eliminating preparation of multiple reagents and reducing the opportunity for technician error. Since there is no need to create a standard curve, create a positive product control and reconstitute LAL, there is no need for a negative control by the user. However, it is recommended that when performing sample dilutions, the diluent be tested for endotoxin prior to use.