0-8mm threaded vacuum stopcock with PTFE plug with UHDPE handle. Needle valve permits smooth valve adjustment down to 0.1 cc/min flow rate. Double PTFE ring seals prevent exposure of FETFE backup O-ring to corrosive gases and liquids.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
0-8mm threaded vacuum stopcock with PTFE plug with UHDPE handle. Needle valve permits smooth valve adjustment down to 0.1 cc/min flow rate. Double PTFE ring seals prevent exposure of FETFE backup O-ring to corrosive gases and liquids.