missing translation for 'onlineSavingsMsg'
Learn More

0.8 Percent Platinum on Carbon Powder, Reduced, Water Wet, 23 microns, BASF Catalyst, Spectrum™ Chemical

Catalog No. p-7161744
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18607909 5 g
18607910 25 g
18607911 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607909 Supplier Spectrum Chemical Mfg Cor Supplier No. PT1105G
Only null left

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
IUPAC Name platinum
SMILES [Pt]

Specifications

Quantity 5 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.