missing translation for 'onlineSavingsMsg'
Learn More
Learn More
0.8 Percent Platinum - 0.3 Percent Molybdenum on Carbon Powder, Reduced, Water Wet, 25 microns, BASF Catalyst, Spectrum™ Chemical
$345.23 - $3827.83
Chemical Identifiers
| CAS | 6-4-7440 |
|---|---|
| Molecular Formula | Pt |
| Molecular Weight (g/mol) | 195.08 |
| MDL Number | MFCD00011179 |
| InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
| IUPAC Name | platinum |
| SMILES | [Pt] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-607-912
|
Spectrum Chemical Mfg Cor
PT2005G |
5 g |
Each for $345.23
|
|
|||||
|
18-607-913
|
Spectrum Chemical Mfg Cor
PT20025G |
25 g |
Each for $1,399.50
|
|
|||||
|
18-607-914
|
Spectrum Chemical Mfg Cor
PT200100G |
100 g |
Each for $3,827.83
|
|
|||||
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.