Learn More
Fine Science Tools Dumont #2 - Laminectomy Forceps, 12cm
Catalog No. NC9067836
Encompass
Style: #2/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.4 x 0.02mm/ Alloy / Material: Inox/ Length: 12cm. These forceps are ideal for breaking off delicate skull bones.
Catalog No. NC9067836 Supplier Fine Science Tools Supplier No. 1122320
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Style: #2/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.4 x 0.02mm/ Alloy / Material: Inox/ Length: 12cm. These forceps are ideal for breaking off delicate skull bones.