Learn More
Thermo Scientific™ Tubes, strips of 8
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.
| Catalog Number | Mfr. No. | Color | Closure Type | Product Type | Quantity | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Closure Type | Product Type | Quantity | Price | Quantity & Availability | ||
|
AB0264
|
Thermo Scientific™
AB0264 |
Natural | Not Included | 8-Strip Tube | 250 Strips |
N/A
|
|
||
|
AB0452
|
Thermo Scientific™
AB0452 |
Natural | Not Included | 8-Strip Tube | 10 Packs of 12 Strips |
N/A
|
|
||
Description
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights
- Compatible with 0.2 mL thermal cycler blocks
- 0.25 mL maximum tube capacity when capped
- Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
Specifications
| Polypropylene |