Get Up to $3,000 in Extra qPCR Products with Purchase
When you buy up to $3,000 of Applied Biosystems™ qPCR consumables, you’ll get a matching amount of qPCR products at no additional cost.
After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
Real-time PCR, also called quantitative or qPCR, is the gold standard for detecting and quantifying nucleic acid targets. Applied Biosystems offers optimized master mixes, assay algorithms, intuitive data analysis software, and smart instrumentation to give you the right tools for your applications. Their products are designed to work together to help create a flexible and efficient qPCR workflow.
Qualifying Products
Applied Biosystems™ Power SYBR™ Green PCR Master Mix 
Offers superior sensitivity and reproducibility, detecting as few as two copies of a target gene over a broad range of template concentrations.
GC-Rich PCR Performance | High |
---|---|
Purity or Quality Grade | LD (Low DNA) |
Concentration | 2X |
For Use With (Application) | Gene Expression |
PCR Method | qPCR |
Detection Method | SYBR |
Polymerase | AmpliTaq Gold DNA Polymerase, AmpliTaq Gold LD |
Product Type | Real Time PCR SYBR Master Mix |
Passive Reference Dye | ROX (Pre-mixed) |
For Use With (Equipment) | 7300 System, 7500 System, 7700 System, 7900HT Fast System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System |
Sample Type | DNA (Genomic), cDNA |
Reaction Speed | Standard |
Label or Dye | SYBR Green |
Product Line | SYBR™ |
Applied Biosystems™ TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG 
TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG is the ideal reagent solution when you need a Master Mix for multiple 5' nuclease DNA applications.
Shipping Condition | Wet Ice |
---|---|
Polymerase | AmpliTaq Gold DNA Polymerase |
GC-Rich PCR Performance | High |
Product Type | Universal PCR Master Mix |
Passive Reference Dye | ROX (Pre-mixed) |
For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
Sample Type | DNA (Genomic), cDNA |
Reaction Speed | Standard |
For Use With (Application) | Gene Expression,Genotyping,miRNA analysis |
PCR Method | qPCR |
Product Line | TaqMan™ |
Detection Method | Primer-probe |
Applied Biosystems™ TaqMan™ Exogenous Internal Positive Control Reagents 
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
TaqMan™ Exogenous Internal Positive Control Reagents contain a pre-optimized internal positive control (IPC) with pre-designed primers and TaqMan™ probe.
GC-Rich PCR Performance | Low |
---|---|
Format | Tube(s) |
Internal Probe Modification | TAMRA™ Quencher (3'), VIC™ (5') |
Product Type | Exogenous Internal Positive Control Reagent |
Reaction Speed | Standard |
Green Features | Sustainable packaging |
PCR Method | qPCR |
Product Line | TaqMan™ |
Applied Biosystems™ Total RNA Control (Human) 
Provided at a convenient reverse-transcription-ready concentration for all RT-PCR reactions
Applied Biosystems™ Eukaryotic 18S rRNA Endogenous Control (FAM™/MGB probe, non-primer limited) 
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Eukaryotic 18S rRNA Endogenous Control (FAM™/MGB probe, non-primer limited)
Shipping Condition | Room Temperature |
---|---|
Content And Storage | Control Primer-Probe Set(s) |
Gene Symbol | 18S |
Green Features | Sustainable packaging |
Amplicon Size | 187 |
Concentration | 20X |
For Use With (Application) | Gene Expression |
PCR Method | 2-step RT-qPCR |
Detection Method | Primer-probe |
Accession Number | X03205.1 |
Target | 18S rRNA |
Format | Tube |
Label or Dye | FAM |
Species | Eukaryotic, Rat, Mouse, Human |
Product Line | FAM™, TaqMan™ |
Applied Biosystems™ TaqMan™ Control Genomic DNA (human) 
Conveniently packaged in a PCR-ready concentration for use as a control template in PCR reactions
Applied Biosystems™ Human ACTB (Beta Actin) Endogenous Control (FAM™/MGB probe, non-primer limited) 
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Human ACTB (Beta Actin) Endogenous Control (FAM™/MGB probe, non-primer limited)
Shipping Condition | Room Temperature |
---|---|
Content And Storage | Control Primer-Probe Set(s) |
Form | Liquid |
Gene Symbol | ACTB |
Green Features | Sustainable packaging |
Amplicon Size | 171 |
Concentration | 20X |
For Use With (Application) | Gene Expression |
PCR Method | 2-step RT-qPCR |
Detection Method | Primer-probe |
Accession Number | NM_001101.2 |
Target | β-actin |
Format | Tube |
Label or Dye | FAM |
Species | Human |
Product Line | FAM™, TaqMan™ |
Applied Biosystems™ Human GUSB (Beta Glucuronidase) Endogenous Control (FAM™/MGB probe, non-primer limited) 
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Human GUSB (Beta Glucuronidase) Endogenous Control (FAM™/MGB probe, non-primer limited)
Shipping Condition | Room Temperature |
---|---|
Content And Storage | Control Primer-Probe Set(s) |
Gene Symbol | GUSB |
Green Features | Sustainable packaging |
Amplicon Size | 81 |
Concentration | 20X |
For Use With (Application) | Gene Expression |
PCR Method | 2-step RT-qPCR |
Detection Method | Primer-probe |
Accession Number | NM_000181.1 |
Target | GUSB |
Format | Tube |
Label or Dye | FAM |
Species | Human |
Product Line | FAM™, TaqMan™ |
Applied Biosystems™ Human GAPD (GAPDH) Endogenous Control (FAM™/MGB probe, non-primer limited) 
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Human GAPD (GAPDH) Endogenous Control (FAM™/MGB probe, non-primer limited)
Shipping Condition | Room Temperature |
---|---|
Content And Storage | Control Primer-Probe Set(s) |
Form | Liquid |
Gene Symbol | GAPDH |
Green Features | Sustainable packaging |
Amplicon Size | 122 |
Concentration | 20X |
For Use With (Application) | Gene Expression |
PCR Method | 2-step RT-qPCR |
Detection Method | Primer-probe |
Accession Number | NM_002046.3 |
Target | GAPDH |
Format | Tube |
Label or Dye | FAM |
Species | Human |
Product Line | FAM™, TaqMan™ |
Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG 
TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.
GC-Rich PCR Performance | High |
---|---|
Fidelity (vs. Taq) | 2 X |
Concentration | 2X |
For Use With (Application) | Gene Expression,Genotyping,miRNA analysis |
PCR Method | qPCR |
Detection Method | Primer-probe |
Polymerase | AmpliTaq DNA Polymerase |
Format | Tube |
Product Type | Fast Universal PCR Master Mix |
Passive Reference Dye | ROX (Pre-mixed) |
For Use With (Equipment) | 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode |
Sample Type | DNA (Genomic), cDNA |
Reaction Speed | Fast |
Product Line | AmpErase™, TaqMan™ |
Applied Biosystems™ Human B2M (Beta-2-Microglobulin) Endogenous Control (FAM™/MGB probe, non-primer limited) 
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Human B2M (Beta-2-Microglobulin) Endogenous Control (FAM™/MGB probe, non-primer limited)
Shipping Condition | Room Temperature |
---|---|
Content And Storage | Control Primer-Probe Set(s) |
Gene Symbol | B2M |
Green Features | Sustainable packaging |
Amplicon Size | 75 |
Concentration | 20X |
For Use With (Application) | Gene Expression |
PCR Method | 2-step RT-qPCR |
Detection Method | Primer-probe |
Accession Number | NM_004048.2 |
Target | B2M |
Format | Tube |
Label or Dye | FAM |
Species | Human |
Product Line | FAM™, TaqMan™ |
Applied Biosystems™ Mouse ACTB (Actin, Beta) Endogenous Control (VIC™/MGB probe, primer limited) 
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Mouse ACTB (Actin, Beta) Endogenous Control (VIC™/MGB probe, primer limited)
Applied Biosystems™ SYBR™ Green Universal Master Mix 
Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.
Polymerase | AmpliTaq Gold DNA Polymerase |
---|---|
GC-Rich PCR Performance | High |
Format | Tube |
Product Type | Real Time PCR SYBR Master Mix |
Passive Reference Dye | ROX (Pre-mixed) |
Reaction Speed | Standard |
Label or Dye | SYBR Green |
Concentration | 2X |
For Use With (Application) | Gene Expression |
PCR Method | qPCR |
Product Line | SYBR™ |
Detection Method | SYBR |
Applied Biosystems™ TaqMan™ GTXpress™ Master Mix 
Combines AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye, and ROX™ dye in a convenient single vial format.
GC-Rich PCR Performance | High |
---|---|
Concentration | 2X |
For Use With (Application) | Genotyping |
PCR Method | qPCR |
Applied Biosystems™ MeltDoctor™ HRM Positive Control Kit 
Provides nucleic acid controls to demonstrate and troubleshoot High Resolution Melt (HRM) analysis