Get 1 Extra Precision Water Bath When You Buy 2
New Lab Start-up Program
When you purchase two identical qualifying Thermo Scientific™ Precision™ Water Baths, you’ll get an extra one of the same bath at no additional cost. View the qualifying products below. After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
Support your applications with Thermo Scientific Precision general purpose water baths, designed to maintain temperatures from ambient to 100°C and available in capacities from 2 L to 28 L.
Get 1 Extra Precision Water Bath When You Buy 2
Redeem Now
Promotion: 223
You must apply and qualify for NLSU to redeem this promotion.
Qualifying Products
    Catalog Number Description Price Quantity & Availability  
Success
TSGP20 Details
Thermo Scientific™ Precision™ General Purpose Baths Capacity (Metric): 20 L
Product Specifications
Capacity (Metric) 20 L
Description 20L General Purpose Water Bath
Model Precision GP 20
Display Monochrome LCD
Material Epoxy Powder-coated Steel
Certifications/Compliance UL, CE
Includes Clear Polycarbonate Gable Cover and Rubber Duck
Electrical Requirements 120/230 V 50/60 Hz
Temperature Uniformity ±0.2°C
Heating Capacity 1200 w
Amperage 9.0-10.5 / 4.3-5.0 (120V/230V)
Dimensions (L x W x H) 15.4 x 21.8 x 9.2 in. (392 x 555 x 233 mm)
Controller Type Digital
Chamber Material Stainless Steel
Temperature Stability ±0.1°C
Temperature Range Ambient to 100°C
Temperature Presets 4
TSGP10 Details
Thermo Scientific™ Precision™ General Purpose Baths Capacity (Metric): 10 L
Product Specifications
Capacity (Metric) 10 L
Description 10L General Purpose Water Bath
Model Precision GP 10
Display Monochrome LCD
Material Epoxy Powder-coated Steel
Certifications/Compliance UL, CE
Includes Clear Polycarbonate Gable Cover and Rubber Duck
Electrical Requirements 120/230 V 50/60 Hz
Temperature Uniformity ±0.2°C
Heating Capacity 800 w
Amperage 6.0-7.0 / 2.8-3.4 (120V/230V)
Dimensions (L x W x H) 15.5 x 15.1 x 9.2 in. (393 x 383 x 233 mm)
Controller Type Digital
Chamber Material Stainless Steel
Temperature Stability ±0.1°C
Temperature Range Ambient to 100°C
Temperature Presets 4
TSGP28 Details
Thermo Scientific™ Precision™ General Purpose Baths Capacity (Metric): 28 L
Product Specifications
Capacity (Metric) 28 L
Description 28L General Purpose Water Bath
Model Precision GP 28
Display Monochrome LCD
Material Epoxy Powder-coated Steel
Certifications/Compliance UL, CE
Includes Clear Polycarbonate Gable Cover and Rubber Duck
Electrical Requirements 120/230 V 50/60 Hz
Temperature Uniformity ±0.2°C
Heating Capacity 1200 w
Amperage 9.0-10.5 / 4.3-5.0 (120V/230V)
Dimensions (L x W x H) 15.4 x 21.8 x 11.1 in. (392 x 555 x 282 mm)
Controller Type Digital
Chamber Material Stainless Steel
Temperature Stability ±0.1°C
Temperature Range Ambient to 100°C
Temperature Presets 4
TSGP02 Details
Thermo Scientific™ Precision™ General Purpose Baths Capacity (Metric): 2 L
Product Specifications
Capacity (Metric) 2 L
Description 2L General Purpose Water Bath
Model Precision GP 02
Display Monochrome LCD
Material Epoxy Powder-coated Steel
Certifications/Compliance UL, CE
Includes Clear Polycarbonate Gable Cover and Rubber Duck
Electrical Requirements 120/230 V 50/60 Hz
Temperature Uniformity ±0.2°C
Heating Capacity 200 w
Amperage 1.5-1.8 / 0.7-0.9 (120V/230V)
Dimensions (L x W x H) 9.1 x 7.8 x 9.2 in. (230 x 199 x 233 mm)
Controller Type Digital
Chamber Material Stainless Steel
Temperature Stability ±0.1°C
Temperature Range Ambient to 90°C
Temperature Presets 4
TSGP05 Details
Thermo Scientific™ Precision™ General Purpose Baths Capacity (Metric): 5 L
Product Specifications
Capacity (Metric) 5 L
Description 5L General Purpose Water Bath
Model Precision GP 05
Display Monochrome LCD
Material Epoxy Powder-coated Steel
Certifications/Compliance UL, CE
Includes Clear Polycarbonate Gable Cover and Rubber Duck
Electrical Requirements 120/230 V 50/60 Hz
Temperature Uniformity ±0.2°C
Heating Capacity 300 w
Amperage 2.2-2.7 / 1.0-1.3 (120V/230V)
Dimensions (L x W x H) 9.7 x 14 x 9.1 in. (246 x 355 x 232 mm)
Controller Type Digital
Chamber Material Stainless Steel
Temperature Stability ±0.1°C
Temperature Range Ambient to 100°C
Temperature Presets 4