Buy 3, Get 1 on Lysis Beads and Matrix Tubes
2017-nlsu-promo-banner
Purchase three MP Biomedicals™ FastPrep™ Lysis Beads and Matrix Tubes for sample disruption and get a fourth at no additional cost.
Features and Benefits
FastPrep Lysing Beads and Matrices effectively disrupt the cell walls of even the toughest samples to produce high yields of nucleic acids and proteins.
Choose the right matrix for your application:
  • Matrix A: All sample types (animal, plant, bacterial, yeasts, fungi)
  • Matrix B: Bacteria, spores
  • Matrix C: Yeast and yeast spores, fungi and fungal spores, algae
  • Matrix D: Animal and plant tissues, cultured cells
  • Matrix E: Soil, feces, environmental water, wastewater, sludge, pollen
105-product-image
nlsu-redeem
Download the redemption form (PDF, 36KB)
Qualifying Products
Restrictions
Offer valid until January 31, 2018.
For Healthcare customers, please note that the value of the specific offer(s) the customer may receive from the manufacturer under this program is a discount or other reduction in price to the customer under section 1128B(b)(3)(A) of the Social Security Act [42 U.S.C. 1320a-7b (b) (3)(a)]. Accordingly, the customer shall disclose this and any other discounts or other reduction in price received under this program under any state or federal program which provides cost or charge-based reimbursements to the customer for the products or services purchased under this program. By participating in this promotion, you warrant that you are not a Health Care Practitioner or Health Care Professional as defined in the Code of Massachusetts Regulations, 105 CMR 970, or a healthcare professional as defined under 18.V.S.A. §4631, or a healthcare professional as defined under any similar federal, state or local law regulation.