accessibility menu, dialog, popup

UserName

H3COCl

CAS RN 75484-32-1

IUPAC Name: 2-(chloromethyl)-2-ethyloxirane
Synonyms: 2-(chloromethyl)-2-ethyloxirane
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: JOBYTHHZLJPOLW-UHFFFAOYNA-N
SMILES: CCC1(CCl)CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results