Get a pH Buffer Kit with pH/mV Meter Purchase
New Lab Start-up Program
When you purchase a qualifying Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter, you’ll get a Fisherbrand accumet Standard All-in-One pH Buffer Kit at no additional cost.
Features and Benefits
Ideal for daily pH testing of your routine laboratory samples, the AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.
Get a pH Buffer Kit with pH/mV Meter Purchase
Redeem Now
Promotion: 229
You must apply and qualify for NLSU to redeem this promotion.
Qualifying Product
    Catalog Number Description Price Quantity & Availability  
Success
13636AB315A Details
Fisherbrandâ„¢ accumetâ„¢ Basic AB315 Benchtop Laboratory pH/mV Meter Type: pH/mV Meter with Probe
Product Specifications
Outputs Port for Computer or Printer
Model AB315
Type pH/mV Meter with Probe
Warranty 3 Years
Material Epoxy Body
Calibration pH, Relative mV, Temperature
Inputs BNC, ATC
Log Function Type Automatic, Manual
Data Points 500
Includes pH/ATC Electrode, Electrode Stand, Power Adapter
Memory Non-volatile
Accuracy (pH) ±0.01
Backlight Option Yes
Voltage 100/240 V
Data Management Details Printed or CSV Format
Frequency 50/60 Hz
Resolution (mV) 1 mV, 0.1 RmV
Description pH/mV Meter with pH/ATC Epoxy-body Double Junction Gel-filled Electrode
Temperature Range (English) 23°F to 221°F
Resolution (Temperature) 0.1°C (0.1°F)
Temperature Range (Metric) -5°C to +105°C
Range (pH) -2 to 18
Certifications/Compliance CE, TUV 3-1, FCC Class A, Without NIST-traceable Calibration Certificate
Stability Indicator Icons
Measurement Modes pH with mV and Temperature, Relative mV with mV and Temperature
Range (mV) ±2000 mV
Calibration Points (pH) USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13
Channels 1
Accuracy (Temperature) ±0.3°C (±0.5°F)
IP Rating IP54
Accuracy (mV) ±0.2 or ±0.05%, Whichever is Greater
Display Type 5 in. LCD
AC Adapter Universal Power Adapter
Temperature Compensation Automatic, Manual
Probe Type Gel-filled pH/ATC Electrode
Calibration Points 1 to 5 (pH), 1 (Relative mV), 1 (Temperature)
Range (Relative mV) ±2000 mV
Resolution (pH) 0.01, 0.1
Redeemable Product
Catalog NumberDescription
Catalog Number
Description
Fisherbrandâ„¢ accumetâ„¢ Standard All-in-One pH Buffer Kit