Get an Extended Warranty with Thermo Scientific Spectrophotometer Purchase
New Lab Start-up Program
When you purchase a qualifying Thermo Scientific NanoDrop Lite Plus Spectrophotometer, you’ll recieve a Unity™ Lab Services extended warranty for it at no additional cost.
Features and Benefits
Quickly and easily assess sample concentration and purity ratios with the entry-level Thermo Scientific NanoDrop Lite Plus Microvolume Spectrophotometer.
Buy a Thermo Scientific Spectrophotometer and Get an Extended Warranty
Redeem Now
Promotion: 222
You must apply and qualify for NLSU to redeem this promotion.
Qualifying Product
    Catalog Number Description Price Quantity & Availability  
Success
NDLPLUSUSCA Details
Thermo Scientific™ NanoDrop Lite Plus Spectrophotometers Includes: ND Lite Plus
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Light Source Xenon Flash
For Use With (Application) Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
Type Microvolume UV-Vis Spectrophotometer
Warranty One year
Absorbance Range 0.04 to 30 Abs (10 mm equivalent)
Material Stainless Steel (303), Quartz Fiber
Weight (Metric) 2.7 kg (without printer)
Wavelength Range 230 nm, 260 nm, 280 nm
Product Line Microvolume
Includes ND Lite Plus
Wavelength Accuracy ± 1 nm
Voltage 100/240 V
System Requirements Standalone control with data export via USB Flash Drive
Processor Standalone control with data export via USB Flash Drive
Height (Metric) 22 cm
No. of Samples 1
Absorbance Accuracy 3% at 0.97 A, 302 nm, 23 ± 2°C Abs/mm at 25 °C
Compatibility Local Control Only
Display LCD Touchscreen
Certifications/Compliance UL/CSA and CE
Detector Type 2048-element CMOS Linear Image Sensor
Concentration 1,500 ng/ÎĽL (1,200 ng/ÎĽL) dsDNA (RNA)45 mg/mL (21 mg/mL) BSA (lgG)
Weight (English) 6.0 lbs (without printer)
Power Consumption 18 W (standby < 3 Watts)
Detection Limits 2.0 ng/ÎĽL (1.6 ng/ÎĽL) dsDNA (RNA)0.06 mg/mL (0.03 mg/mL) BSA (lgG)
Sample Volume (Metric) 1 to 2 ÎĽL
Spectral Resolution ≤ 1.8 nm (FWHM at Hg 254 nm)
Depth (Metric) 22 cm
Measurement Time ≤ 5 seconds
Pathlength 1.0 mm and 0.2 mm (auto adjust)
Width (Metric) 27 cm
Printer Included No
Connections USB 2.0 Port
Footprint 27 x 22 x 22 cm (0.6 x 8.7 x 8.7 in); W x D X H
Redeemable Product
Catalog NumberDescription
Catalog Number
Description
Unity™ Lab Services Extended Warranty Thermo Scientific™ NanoDrop™ Lite Spectrophotometer