Get Corning Equipment with $1,000 Purchase
When you buy $1,000 in qualifying Corning™, Falcon, or Axygen products, you’ll get a Corning™ Stripettor™ Ultra Pipet Controller or Corning™ LSE™ Mini Microcentrifuge at no additional cost. To take advantage of this offer, order using the catalog numbers listed, log in to the Redemption Center, and complete the redemption form to receive your pipet controller or microcentrifuge.
Features and Benefits
Advance your research with the valuable, high-quality instruments that have complemented Corning plasticware and glassware for more than 50 years. Labs worldwide count on Corning to provide innovative scientific laboratory equipment for life sciences, chemistry, and industrial applications.
Corning™ Lambda Plus Single-Channel Pipettor, Adjustable, Autoclavable
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Designed for universal fit with all common brands of pipet tips
Includes | Key is included with each pipette for easy in-house calibration |
---|---|
Type | Single-Channel Pipettor |
Warranty | 3 Years |
Number of Channels | 1 |
Certifications/Compliance | ISO8655 |
UV Monitoring | Yes |
Autoclavable | Autoclavable |
Product Line | Lambda Plus |
Corning™ Lambda Plus 8 and 12-Channel Pipettors
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Designed for universal fit with all common brands of pipet tips
Type | Pipette |
---|---|
Warranty | 3 Years |
For Use With | Universal Pipette Tip |
Certifications/Compliance | CE |
UV Monitoring | Yes |
Autoclavable | Autoclavable |
Sterility | Non-sterile |
Product Line | Lambda Plus |
Corning™ Lambda™ EliteTouch™ Single-Channel Pipettors, Autoclavable
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Corning™ Lambda™ EliteTouch™ Single-Channel Pipettors have been engineered to provide the highest levels of ergonomics, accuracy, and precision.
Includes | Instruction manual, QC Certificate, In-lab calibration tool, 3 color pushbuttons |
---|---|
Grip Type | Ergonomic |
Finger Style | Ergonomic |
Type | Pipette |
Warranty | 3 Years |
Number of Channels | 1 |
Quantity | 1 |
Certifications/Compliance | ISO8655 |
Autoclavable | Autoclavable |
Product Line | Lambda EliteTouch |
Corning™ Lambda™ EliteTouch™ 8 Multi-Channel Pipettors, Autoclavable
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Corning™ Lambda™ EliteTouch™ 8 Multi-Channel Pipettors have been engineered to provide the highest levels of ergonomics, accuracy, and precision.
Includes | Instruction manual, QC Certificate, In-lab calibration tool, 3 color pushbuttons |
---|---|
Grip Type | Ergonomic |
Finger Style | Ergonomic |
Type | Pipette |
Warranty | 3 Years |
Number of Channels | 8 |
Quantity | 1 |
Certifications/Compliance | ISO8655 |
Autoclavable | Autoclavable |
Product Line | Lambda EliteTouch |
Corning™ Lambda™ EliteTouch™ Pipettor Starter Kit
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Corning™ Lambda™ EliteTouch™ Pipettor Starter Kit has been engineered to provide the highest levels of ergonomics, accuracy, and precision.
Corning™ LSE™ Compact Centrifuge
Space saving centrifuge ideal for use in life science and industrial research labs
Corning™ LSE™ Shaking Incubators
Cabinet-style incubator has compact footprint and can be stacked to maximize space efficiency. Built in orbital shaker accepts a large assortment of platforms and clamps that can accommodate flasks, Petri dishes, and test tube racks.
Timer Range | 0 to 99 min. |
---|---|
Orbit | 19 mm |
Product Type | Incubator |
Style | Cabinet |
Material | Exterior: Cold rolled steel; Interior: Mirrored stainless steel |
Display | Digital |
Corning™ LSE™ Multiposition Digital Vortexer
Corning™ LSE Multipostion Digital Vortexer is a multipurpose shaker dedicated to a variety of aggressive shaking and mixing applications.
Shipping Weight (Metric) | 7.3 kg |
---|---|
Load Bearing Capacity (English) | 4.4 lb. |
Load Bearing Capacity (Metric) | 2 kg |
Description | Requires a platform |
Timer Range | 0 to 99 minutes or continuous |
Product Type | Vortexer |
Warranty and Service Offering | 5 Years |
Frequency | 50/60 Hz |
For Use With (Application) | General Purpose Mixing |
Shipping Weight (English) | 16 lb. |
Temperature Range (Metric) | 4°C to 65°C |
Speed Range | 100 to 1200 rpm |
Corning™ LSE™ Benchtop Shaking Incubator with Platform
The Corning benchtop shaking incubator has low pivot point which provides space-saving size sure to fit on lab bench and under shelves.
Height (Metric) | 40 cm |
---|---|
Depth (English) | 20.8 in. |
Temperature Accuracy | Temperature: ±0.5°C |
Orbit | 19 mm |
Width (Metric) | 37 cm |
Dimensions (LxW) Platform | 30 x 30 cm (11.8 x 11.8 in.) |
Timer Range | 1 min. to 99 hr., Continuous |
Product Type | Incubator |
Depth (Metric) | 53 cm |
Style | Benchtop |
Weight (English) | 43 lb. |
Temperature Range (Metric) | Ambient +5° to 70°C |
Speed Range | 20 to 300 rpm |
Weight (Metric) | 19.5 kg |
Height (English) | 15.7 in. |
Corning™ Lambda Plus Pipettor Starter Kit
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes
Corning™ LSE™ Digital Microplate Shaker
Features digital control of shaking speed and run time
Corning™ LSE™ Vortex Mixer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Variable speed mixer provides fast, efficient mixing with minimal vibration
Corning™ Lambda™ EliteTouch™ 12 Multi-Channel Pipettors, Autoclavable
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More
Corning™ Lambda™ EliteTouch™ 12 Multi-Channel Pipettors have been engineered to provide the highest levels of ergonomics, accuracy, and precision.
Includes | Instruction manual, QC Certificate, In-lab calibration tool, 3 color pushbuttons |
---|---|
Grip Type | Ergonomic |
Finger Style | Ergonomic |
Type | Pipette |
Warranty | 3 Years |
Number of Channels | 12 |
Quantity | 1 |
Certifications/Compliance | ISO8655 |
Autoclavable | Autoclavable |
Product Line | Lambda EliteTouch |
Corning™ Step-R™ Repeating Pipettor
The Corning™ Step-R™ Repeating Pipettor is ideal for dispensing volatile, dense, viscous or foaming liquids, and for all applications where long dosing series are required
Corning™ LSE™ Digital Water Bath, 6L
The Corning LSE Digital Water Bath is a personal-sized bath with seamless chamber that can accommodate up to 6L of water or can serve as dry bath when using thermal beads.
Height (Metric) | 23 cm |
---|---|
Description | Water Bath |
Width (English) | 14.9 in. |
Width (English) Interior | Chamber: 12.6 in. |
Dimensions (D x W x H) Interior | 17 x 32 x 18 cm (6.7 x 12.6 x 6.9 in.) |
Capacity (Metric) | 6 L |
Controller Type | PID |
Width (Metric) | 38 cm |
Width (Metric) Interior | 32 cm Chamber |
Depth (Metric) Interior | 17 cm Chamber |
Depth (Metric) | 29 cm |
Temperature Range | Ambient +5° to 99°C |
Weight (English) | 19 lb. |
Depth (English) Interior | Chamber: 6.7 in. |
Weight (Metric) | 8.6 kg |
Height (English) Interior | Chamber: 6.9 in. |
Height (English) | 9.0 in. |
Display | Digital LED |
Catalog Number | Description | |||
---|---|---|---|---|
Catalog Number |
Description Corning Stripettor Ultra Pipet Controller with Universal Power Supply | Learn More | ||
Catalog Number |
Description Corning LSE Mini Microcentrifuge, 100 to 240V | Learn More |