Get Fisherbrand Bead Mill Tubes with Homogenizer Purchase
When you purchase a Fisherbrand™ Bead Mill 24 Homogenizer, you’ll receive 500 Fisherbrand Hard Tissue Pre-Filled Tubes at no additional cost.
After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
The Fisherbrand Bead Mill 24 Homogenizer is used for applications that require grinding, lysing, and homogenizing biological samples before molecular extraction. It can simultaneously process 24 x 2 mL or 0.5 mL tubes.
Fisherbrand Hard Tissue Homogenizing Mix Tubes have a 2 mL capacity and contain 2.8 mm ceramic beads for processing brain, muscle, and other tough sample materials.
| Catalog Number | Description | ||
|---|---|---|---|
Fisherbrand Hard Tissue Homogenizing Mix Tubes, Contain 2.8 mm Ceramic Beads, 2 mL, 50/Pack | Learn More |