Get a Saddle Stool with Biological Safety Cabinet Purchase
When you buy a qualifying Thermo Scientific™ 1300 Series Class II, Type A2 Biological Safety Cabinet (BSC) Package, you’ll get a Thermo Scientific™ Ergolign™ Saddle Stool, a value of over $800, at no additional cost. To take advantage of this offer, order using the catalog numbers listed below, go to the Redemption Center, and complete the form to receive your saddle stool.
May be redeemed on up to four qualifying Thermo Scientific biological safety cabinets purchased on the same order.
Features and Benefits
Thermo Scientific BSCs are certified to protect researchers from contaminated or potentially contaminated materials and samples. Designed with Thermo Scientific™ SmartFlow™ technology, they feature dual-DC motors that automatically balance the cabinet inflow and downflow velocities in real time, even as the filters load. For added confidence, the Digital Airflow Verification (DAVe) signals if conditions exceed predetermined limits.
| Catalog Number | Description | ||
|---|---|---|---|
Thermo Scientific Ergolign Saddle Stool | Learn More |