Get a Glassware Kit with Labconco Freeze Dryer Purchase
When you buy a Labconco™ FreeZone™ Freeze Dryer, you’ll get a freeze dry glassware kit at no additional cost. View the qualifying products below. Once you’ve made your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
Choose FreeZone Freeze Dryers in benchtop and console models that range in size from 2.5 to 18 liters. Find –50°C models for aqueous samples, –84°C models for low eutectic temperatures, and –105°C models for samples containing dilute alcohol. The freeze dry glassware kit includes 300 and 600 mL Labconco™ Fast-Freeze™ Flasks and two stainless-steel adapters.
| Catalog Number | Description | ||
|---|---|---|---|
Labconco™ Freeze-Dry Glassware Kit | Learn More |