Buy a Mastercycler Nexus Thermal Cycler, Get 5 Cases of PCR Consumables
Purchase an Eppendorfâ„¢ Mastercyclerâ„¢ Nexus Thermal Cycler and get five cases of Eppendorfâ„¢ PCR consumables at no extra cost.
Complimentary Products: E0030124260, E0030124286, E0030124812, E0030133374, E951020401
Features and Benefits
The Mastercycler Nexus Thermal Cycler features a universal thermoblock (fits 0.2mL PCR tubes, 96-well PCR plates or 0.5mL PCR tubes); an alternate silver thermoblock allows for faster processing of 0.2mL PCR tubes or 96-well PCR plates. An adjustable-height lid accommodates all PCR consumables.
Eppendorf offers PCR consumables that include 96-well and 384-well PCR plates (for high and medium throughput), divisible plates, PCR tubes and tube strips (for lower throughput), plus sealing options, racks and other accessories.
Qualifying Products
Catalog Number | Description | Price | Quantity & Availability |
---|
Restrictions
Offer valid until January 31, 2018.
For Healthcare customers, please note that the value of the specific offer(s) the customer may receive from the manufacturer under this program is a discount or other reduction in price to the customer under section 1128B(b)(3)(A) of the Social Security Act [42 U.S.C. 1320a-7b (b) (3)(a)]. Accordingly, the customer shall disclose this and any other discounts or other reduction in price received under this program under any state or federal program which provides cost or charge-based reimbursements to the customer for the products or services purchased under this program. By participating in this promotion, you warrant that you are not a Health Care Practitioner or Health Care Professional as defined in the Code of Massachusetts Regulations, 105 CMR 970, or a healthcare professional as defined under 18.V.S.A. §4631, or a healthcare professional as defined under any similar federal, state or local law regulation.