DuPont Lab Safety Products
DuPont Tyvek Coveralls
DuPont™ Tyvek™ full-length coveralls are cut generously for wearing over clothing and feature DuPont exclusive comfort fit design. They are economical enough to discard after wearing, yet strong enough to withstand several wearings. All coveralls are white and include a nylon-zippered front.
Cleanroom
The Fisher Scientific channel is a leading provider of controlled environments and production — from cleanroom, scientific and occupational safety products and services.
Coveralls with Open Wrists and Ankles
Includes a lay-down collar
Coveralls with Elasticized Wrists and Ankles
Features a lay-down collar and a tight, elasticized fit at wrists and ankles
Coveralls with Elasticized Wrists and Ankles Plus Attached Hood
Includes an attached respirator-fit hood with elastic around the face opening, designed to cover the neck and chin and fit around the respirator face mask. A longer zipper extends to the chin for complete neck coverage.
Coveralls with Elasticized Wrists and Ankles and Attached Hood and Boots
Includes an attached respirator-fit hood with elastic around the face opening, designed to cover the neck and chin and fit around the respirator face mask. A longer zipper extends to the chin for complete neck coverage. Attached Tyvek FC books are specially-coated for added skid resistance.
DuPont Tyvek Frocks and Lab Coats
DuPont™ Tyvek™ frocks and lab coats are disposable, yet strong enough to withstand several wearings. They provide excellent protection against submicron particles. All lab coats are white and made from lightweight and tear-resistant Tyvek fabric. Each coat is chemically and biologically inert and provides limited splash protection.
Frocks with Open Wrists
Features a body-length jacket design with front snap closures for easy donning and doffing. They are ideal for applications that do not require the full-body coverage of a coverall or lay-down collar.
Frocks with Elasticized Wrists
Features a knee-length body design with front snap closures for easy donning and doffing. Ideal for applications that do not require full-body coverall coverage, these frocks feature elasticized wrist openings for a tighter fit.
Lab Coats with Open Wrists
Features enhanced frontal protection with a mandarin collar that is designed to fit tightly around a hooded PAPR. This labcoat also includes two front pockets and front snap closures.
DuPont Tyvek Shoe and Boot Covers
DuPont™ Tyvek™ shoe and boot covers protect your feet from dirt, grease and splashes.
Abrasion- and tear-resistant, lightweight and comfortable, Tyvek combines a superior barrier with durability and breathability.
DuPont Tyvek Bib Aprons and Sleeves
DuPont™ Tyvek™ Bib Aprons and Sleeves protect against many hazardous powders. Nonwoven Tyvek fabric helps resist penetration by particles and other contaminants. An inherent barrier stops submicron particles even after abrasion.