Labconco FreeZone Benchtop Freeze Dryers
Labconco™ FreeZone™ Benchtop Freeze Dryers offer real-time on-screen monitoring of sample data via an easy-to-use touch screen display. They are designed for lyophilizing light to moderate sample loads, and they are compact, making them ideal for labs with limited floor space.
The Labconco™ Lyo-Works™ Operating System in your FreeZone Freeze Dryer is more than a large touch screen display. It’s a set of powerful controls and useful insights about your freeze drying process. The Lyo-Works OS lets you stay connected from anywhere with real-time monitoring of sample data and status update emails. The software offers data and protocol program storage and data transfer via ethernet and USB port, so no additional recorders or computers are necessary.