Gilson PIPETMAN M Connected Pipettes
With Gilsonâ„¢ PIPETMANâ„¢ M Connected Bluetoothâ„¢-enabled electronic pipettes, you can expect accuracy and precision from both standard and repetitive pipetting modes. Requiring virtually no pipetting forces to aspirate and dispense samples, PIPETMAN M Connected pipettes may help reduce pipetting fatigue and increase efficiency.
They also make planning and executing protocols faster, easier, and more reliable through real-time interaction with the Gilson™ TRACKMAN™ Connected kit. Choose from 20 PIPETMAN M Connected pipette models ranging in volume from 0.5 µL to 10 mL for single-channel varieties and 0.5 µL to 1,200 µL for multichannel styles.
Catalog Number | Description | Price | Quantity & Availability |
---|